Learn More
Hach Company Digital ORP Sensor PEEK Conv.
Catalog No. 5000207787 Shop All Hach Company Products
Encompass
Digital ORP Sensor-PEEK-Convertible-Non-Hazardous EnvironmentGeneral Purpose Online Process ORP Sensor with Integrated Digital Electronics for in Plug and Playin with Hach sc Digital Controllers - Platinum ORP Electrode PEEK Housing Convertible Mount 10m Cable
Catalog No. 50-002-07787 Supplier Hach Company Supplier No. DRD1P5.1
Digital ORP Sensor-PEEK-Convertible-Non-Hazardous EnvironmentGeneral Purpose Online Process ORP Sensor with Integrated Digital Electronics for in Plug and Playin with Hach sc Digital Controllers - Platinum ORP Electrode PEEK Housing Convertible Mount 10m Cable