Learn More
Hach Company Digital ORP Sensor SS Immers.
Catalog No. 5000207786 Shop All Hach Company Products
Encompass
Digital ORP Sensor-Stainless Steel-Immersion-Non-Hazardous EnvironmentGeneral Purpose Online Process ORP Sensor with Integrated Digital Electronics for in Plug and Playin with Hach sc Digital Controllers - Platinum ORP Electrode Stainless Housing Immersion Mount 10m Cable
Catalog No. 50-002-07786 Supplier Hach Company Supplier No. DRS5.1
Digital ORP Sensor-Stainless Steel-Immersion-Non-Hazardous EnvironmentGeneral Purpose Online Process ORP Sensor with Integrated Digital Electronics for in Plug and Playin with Hach sc Digital Controllers - Platinum ORP Electrode Stainless Housing Immersion Mount 10m Cable