Learn More
Chemglass Life Sciences Digital Overhead Stirrer with Timer, 115V
SDP
Catalog No. 501311195
Encompass_Preferred
Digital Overhead Stirrer with Timer, 115V. The digital display of this top performing stirrer motor provides easy-to-read details: set speed, actual speed, torque and time. Operating time can be set up to 999 minutes and 59 seconds. After the end of the set time, stirring stops automatically.Suitable for all medium to high-viscosity mixing tasks this overhead stirrer motor boasts a maximum viscosity of 50,000 mPa/centipoise. The speed is continuously adjustable from 50 to 2000 rpms with a maximum volume of 40 liters of water. Torque compensation ensures that exact speed under changing load is ensured.The technopolymer case is light-weight, chemical-resistant and rustproof. It’s also sealed to prevent internal corrosion and penetration by aggressive liquids and harmful vapors. Internally, aluminum components support a robust electric stirrer motor built to provide years of reliable service. Safety is built-in with overload, over current and over temperature protection.
Catalog No. 50-131-1195 Supplier Chemglass Life Sciences Supplier No. CG202410
Digital Overhead Stirrer with Timer, 115V. The digital display of this top performing stirrer motor provides easy-to-read details: set speed, actual speed, torque and time. Operating time can be set up to 999 minutes and 59 seconds. After the end of the set time, stirring stops automatically.Suitable for all medium to high-viscosity mixing tasks this overhead stirrer motor boasts a maximum viscosity of 50,000 mPa/centipoise. The speed is continuously adjustable from 50 to 2000 rpms with a maximum volume of 40 liters of water. Torque compensation ensures that exact speed under changing load is ensured.The technopolymer case is light-weight, chemical-resistant and rustproof. It’s also sealed to prevent internal corrosion and penetration by aggressive liquids and harmful vapors. Internally, aluminum components support a robust electric stirrer motor built to provide years of reliable service. Safety is built-in with overload, over current and over temperature protection.