Learn More
Medicus Health Digital Paraffin Wax Trimmer with Magnetic Drip Tray
SDP
Catalog No. NC3666261
Encompass
Digital Paraffin Wax Trimmer with Magnetic Drip Tray provides an efficient solution for removing excess paraffin from tissue embedding cassettes, for enhanced slicing precision in a microtome.Digital Paraffin Wax Trimmer with Magnetic Drip Tray is designed to simplify wax disposal and prevent messy paraffin shavingsHeated, grooved surfaces allow quick and easy removal of extra paraffin allowing the melted wax to drain cleanly into a removable catch basinSimple controls: LED Display and Rotary Knob for easy temperature adjustmentLarge working surface allows multiple tissue cassettes to be trimmed at a timeMagnetically attached drip trayComes with 5 disposable tray liners for easy collection and disposal of waxContinuously adjustable between room temperature and 99°CReaches the set maximum temperature within 5 to 10 minutes, with a temperature accuracy of ±1°CPower failure recovery mode and over-temperature protectionSupports co
Catalog No. NC3666261 Supplier Medicus Health Supplier No. 7515M2
Digital Paraffin Wax Trimmer with Magnetic Drip Tray provides an efficient solution for removing excess paraffin from tissue embedding cassettes, for enhanced slicing precision in a microtome.Digital Paraffin Wax Trimmer with Magnetic Drip Tray is designed to simplify wax disposal and prevent messy paraffin shavingsHeated, grooved surfaces allow quick and easy removal of extra paraffin allowing the melted wax to drain cleanly into a removable catch basinSimple controls: LED Display and Rotary Knob for easy temperature adjustmentLarge working surface allows multiple tissue cassettes to be trimmed at a timeMagnetically attached drip trayComes with 5 disposable tray liners for easy collection and disposal of waxContinuously adjustable between room temperature and 99°CReaches the set maximum temperature within 5 to 10 minutes, with a temperature accuracy of ±1°CPower failure recovery mode and over-temperature protectionSupports co