Learn More
Meridian Life Science Digital PCR Mix 5x Not, 40ML
SDP
Catalog No. 5000856290
Encompass_Preferred
Digital PCR Mix, 5x combines state-of-the-art buffer chemistry, PCR enhancers and stabilizers, along with antibody-mediated hot-start polymerase, dNTPs, and MgCl2. Specifically designed for rapid thermal cycling conditions. Digital PCR Mix, 5x delivers highly reproducible and accurate assays, providing excellent results in multiplex assays.
Catalog No. 50-008-56290 Supplier Meridian Life Science Supplier No. MDX07440
Digital PCR Mix, 5x combines state-of-the-art buffer chemistry, PCR enhancers and stabilizers, along with antibody-mediated hot-start polymerase, dNTPs, and MgCl2. Specifically designed for rapid thermal cycling conditions. Digital PCR Mix, 5x delivers highly reproducible and accurate assays, providing excellent results in multiplex assays.