Learn More
Hach Company Digital pH Sensor PEEK Conv.
Catalog No. 5000207791 Shop All Hach Company Products
Encompass
Digital pH Sensor-PEEK-Convertible-Non-Hazardous EnvironmentHach s patented differential electrode measurement technique ensures more accurate pH readings providing increased reliability to reduce downtime and maintenance for measuring pH over traditional pH sensors Our DPD1P1 digital pH probe is Convertible Mount complete with a 10m cable and compatible with Hach SC controllers This instrument connects to Claros Hach s innovative Water Intelligence System enabling you to seamlessly connect and manage instruments data and process anywhere anytime The result is greater confidence in your data and improved efficiency in your operations To unlock the full potential of Claros insist on Claros Enabled instruments
Catalog No. 50-002-07791 Supplier Hach Company Supplier No. DPD1P1.1
Digital pH Sensor-PEEK-Convertible-Non-Hazardous EnvironmentHach s patented differential electrode measurement technique ensures more accurate pH readings providing increased reliability to reduce downtime and maintenance for measuring pH over traditional pH sensors Our DPD1P1 digital pH probe is Convertible Mount complete with a 10m cable and compatible with Hach SC controllers This instrument connects to Claros Hach s innovative Water Intelligence System enabling you to seamlessly connect and manage instruments data and process anywhere anytime The result is greater confidence in your data and improved efficiency in your operations To unlock the full potential of Claros insist on Claros Enabled instruments