Learn More
Research Products International Corp Nichipet EX Digital Pipettor, 0.1 - 2.0ul
SDP
Supplier:  Research Products International Corp 249800
Encompass
The Nichipet EX II Digital Pipettor offers the best value in a high performance laboratory pipettor. There simply is not a comparable product that matches the features and performance of this instrument. The entire unit can be steam sterilized at 121°C for up to twenty minutes. Enhanced UV resistance allows repeated exposure to ultraviolet rays without affecting durability or accuracy. Additional benefits include: ergonomic grip surface; lightweight design; easy to set dial indicator with locking lever; integral tip ejector; slim barrel style; calibration certificate and twelve month warranty.
Catalog No. 50-998-522