Learn More
Cole-Parmer Cole-Parmer Digital Pressure Gauge, 0 to 145 psi, with 1/4" NPT(M) Connections
Catalog No. NC1568361 Shop All Cole-Parmer Products
Encompass_Preferred
Designed for use in numerous applications and industries, these digital gauges can be used for air pressure measurement in marine and diving, filtration, compressor and pneumatic regulators measurement, laboratory vacuum and pressure measurement, as well as aviation and defense, and general OEM use where air or compatible gases are used and brass can be used.
These high-accuracy, low-cost digital gauges have a four-digit, 1/2"H LCD and read in psi, bar, mPa, and kg/cm2. Gauges have a 1 3/4" (44.5 mm) dial and come in three styles — 0 to 145 psi in 1/8" or 1/4" NPT(M) connection, and vacuum to 100 psi in 1/8" NPT(M) connection.
Catalog No. NC1568361 Supplier Cole-Parmer Supplier No. 6895035
Designed for use in numerous applications and industries, these digital gauges can be used for air pressure measurement in marine and diving, filtration, compressor and pneumatic regulators measurement, laboratory vacuum and pressure measurement, as well as aviation and defense, and general OEM use where air or compatible gases are used and brass can be used.
These high-accuracy, low-cost digital gauges have a four-digit, 1/2"H LCD and read in psi, bar, mPa, and kg/cm2. Gauges have a 1 3/4" (44.5 mm) dial and come in three styles — 0 to 145 psi in 1/8" or 1/4" NPT(M) connection, and vacuum to 100 psi in 1/8" NPT(M) connection.