Learn More
Bellco Glass, Inc. Digital Rocker Complete, 230V
SDP
Catalog No. 501946123
Encompass_Preferred
The Bellco Digital Rocker is a slow speed rocker for general laboratorycell culture experimentation. This Bellco design incorporates a rockingplatform that accepts a variety of optional trays. Eight tray models areavailable, including three equipped with flask clamps for common sizesof flasks. Peg and Grommet style trays make changing convenient. Thetimer circuit permits automatic shutoff after 3-120 minutes.. MADE IN THE USA
Catalog No. 50-194-6123 Supplier Bellco Glass, Inc. Supplier No. 774310230
The Bellco Digital Rocker is a slow speed rocker for general laboratorycell culture experimentation. This Bellco design incorporates a rockingplatform that accepts a variety of optional trays. Eight tray models areavailable, including three equipped with flask clamps for common sizesof flasks. Peg and Grommet style trays make changing convenient. Thetimer circuit permits automatic shutoff after 3-120 minutes.. MADE IN THE USA