Learn More
Bellco Glass, Inc. Digital Shaker,115v W/Power Supply - No Tray
SDP
Catalog No. 501945962
Encompass_Preferred
The Bellco Digital Shaker is a compact, heavy-duty variable speed shaker. It is used for the agitation of electrophoretic gels and filter assay media for staining, destaining and assay development. The Shaker accommodates a variety of vessels. It is continuously adjustable from 30 to 350 RPM. This Bellco design incorporates an orbital platform that accepts a variety of optional trays. A variety of tray models are available, including three equipped with flask clamps for common sizes of flasks. Peg and Grommet style trays make changing convenient. The timer circuit permits automatic shutoff after 3-120 minutes. MADE IN THE USA
Catalog No. 50-194-5962 Supplier Bellco Glass, Inc. Supplier No. 764410115
The Bellco Digital Shaker is a compact, heavy-duty variable speed shaker. It is used for the agitation of electrophoretic gels and filter assay media for staining, destaining and assay development. The Shaker accommodates a variety of vessels. It is continuously adjustable from 30 to 350 RPM. This Bellco design incorporates an orbital platform that accepts a variety of optional trays. A variety of tray models are available, including three equipped with flask clamps for common sizes of flasks. Peg and Grommet style trays make changing convenient. The timer circuit permits automatic shutoff after 3-120 minutes. MADE IN THE USA