Learn More
Electron Microscopy Sciences uSCOPE™ HXII-20 Slide Scanner, 20x (0.40NA) Objective
SDP
Catalog No. 501928302
Encompass
The uSCOPE HXII Digital Microscope Slide Scanner is a low-cost, portable, personal microscope slide scanner that can browse or scan your prepared glass slides. It is completely self-contained, highly portable and easy to setup on the road. The industry-standard USB interface makes it simple to plug in and start capturing images.
The uSCOPE Navigator Software simplifies capturing an area of interest from your glass slides. User-defined filter sequences enable you to apply multi-step image enhancement filters (brightness, contrast, look-up tables, gamma control, sharpness, smoothing, and flat field correction) to captured images or image sets. Deep zoom image sets, which allow you to zoom in and out and navigate large slide scans, are easily created.
Objective: 20x (0.40NA)
@ 960 x 540: ≈ 0.50µm/pixel
@ 1920 x 1080: ≈0.25 µm/pixel
Catalog No. 50-192-8302 Supplier Electron Microscopy Sciences Supplier No. 8901201
Add to Cart
Add to Cart
Request a Quote
The uSCOPE HXII Digital Microscope Slide Scanner is a low-cost, portable, personal microscope slide scanner that can browse or scan your prepared glass slides. It is completely self-contained, highly portable and easy to setup on the road. The industry-standard USB interface makes it simple to plug in and start capturing images.
The uSCOPE Navigator Software simplifies capturing an area of interest from your glass slides. User-defined filter sequences enable you to apply multi-step image enhancement filters (brightness, contrast, look-up tables, gamma control, sharpness, smoothing, and flat field correction) to captured images or image sets. Deep zoom image sets, which allow you to zoom in and out and navigate large slide scans, are easily created.
Objective: 20x (0.40NA)
@ 960 x 540: ≈ 0.50µm/pixel
@ 1920 x 1080: ≈0.25 µm/pixel