Learn More
World Precision Instrument WPI Standard Digital Mouse Stereotaxic Instrument, Non-Rupture Ear Bars
SDP
Catalog No. 502539071
Encompass
Digital Mouse Stereotaxic Instrument, Non-Rupture Ear Bars. A digital mouse-only version of WPI's Standard Stereotaxic Instrument, Non-Rupture Ear Bars paired with the Elevated Platform for Mouse and a mouse-specific bite bar for use on the 'U' frame stereotaxic configuration.  
View our Stereotaxic Frame Compatibility Chart
Catalog No. 50-253-9071 Supplier World Precision Instrument Supplier No. 505389
Add to Cart
Add to Cart
Request a Quote
Digital Mouse Stereotaxic Instrument, Non-Rupture Ear Bars. A digital mouse-only version of WPI's Standard Stereotaxic Instrument, Non-Rupture Ear Bars paired with the Elevated Platform for Mouse and a mouse-specific bite bar for use on the 'U' frame stereotaxic configuration.  
View our Stereotaxic Frame Compatibility Chart