Learn More
Chemglass Life Sciences Digital Motor Controller Only For 1/4HP Explosion Proof Motor, 115V
SDP
Catalog No. 50401821
Encompass_Preferred
Digital Motor Controller Only For Explosion Proof Motor, 115V. Explosion proof (XP) motor features a 5:1 right angle gearbox to reduce the overall height above support frame to 10.5¿. Motor is supplied with a 30ft 18/3 SJEOO cord with twist lock connector. XP motor must be used in conjunction with our digital motor controller, and mounts to all our support frames via the supplied mounting bracket. PLEASE NOTE: A LICENSED ELECTRICIAN SHOULD BE CONSULTED FOR ALL FINAL CONNECTIONS. DIGITAL MOTOR CONTROLLER IS NOT EXPLOSION PROOF AND MUST BE MOUNTED OUTSIDE THE HOOD/XP ENVIRONMENT.
Catalog No. 50-401-821 Supplier Chemglass Life Sciences Supplier No. CG2033X31
Digital Motor Controller Only For Explosion Proof Motor, 115V. Explosion proof (XP) motor features a 5:1 right angle gearbox to reduce the overall height above support frame to 10.5¿. Motor is supplied with a 30ft 18/3 SJEOO cord with twist lock connector. XP motor must be used in conjunction with our digital motor controller, and mounts to all our support frames via the supplied mounting bracket. PLEASE NOTE: A LICENSED ELECTRICIAN SHOULD BE CONSULTED FOR ALL FINAL CONNECTIONS. DIGITAL MOTOR CONTROLLER IS NOT EXPLOSION PROOF AND MUST BE MOUNTED OUTSIDE THE HOOD/XP ENVIRONMENT.