Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LaMotte™ Digital Tap Water Tour
SDPSCI_ED

Catalog No. S44607 Shop All LaMotte Company Products
Change view
Click to view available options
Class Size:
50 Students
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S44607 50 Students
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S44607 Supplier LaMotte™ Supplier No. 597201
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A classroom investigation of how and why the characteristics of tap water can vary and affect everyday activities.

  • After reviewing background material about pH, chlorine, hardness, copper and iron, students work independently and in teams to test their own tap water samples with TesTab™ reagents
  • Students use observations and analyze shared data from the hands-on activities to explore water quality
  • Incorporates math and science activities, which can be modified to fit schedules and learning opportunities
  • Data sheets and puzzles included
  • Great for inclusion classrooms

Specifications

Grade K to 12
Includes Activities, Reagent Tablets, Data Sheets, Puzzles
Class Size 50 Students
Product Type Curriculum and Hands-on Water Chemistry Tests
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.