Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LaMotte™ Digital Tapwater Tour Kit
SDPSCI_ED

Catalog No. S72762 Shop All LaMotte Company Products
Change view
Click to view available options
Includes:
Intructor Manual, Student Manual, TesTab™ reagents and sample bags for 50 students
TesTab™ reagents and sample bags for 50 students
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
S72762 TesTab™ reagents and sample bags for 50 students
S72761 Intructor Manual, Student Manual, TesTab™ reagents and sample bags for 50 students
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S72762 Supplier LaMotte™ Supplier No. R5972

missing translation for 'validMainframeMsg'

A classroom investigation of how and why the characteristics of tap water can vary and affect every day activities

  • Instructor manual including background information, set up and shopping list
  • Student Manual including test procedures, data sheets and puzzles
  • Students work in groups and individually
  • Works on computers or tablets
  • Great for inclusion classrooms
  • Refill includes reagents and sample bags for 50 students

Specifications

Grade 4 to 8
Age 9 to 14
Includes TesTab™ reagents and sample bags for 50 students
Material Plastic sample bags, tableted TesTab™ reagents
Quantity 1
Class Size 50 Students
Product Type Hands-on Water Chemistry Tests
Description Digital Tapwater Tour Kit Refill
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.