Learn More
Chemglass Life Sciences Temperature Monitor Only, Digital, Type "T", 220V, -200 to 250°C, with Electrical Plug for China
SDP
Catalog No. 501531622
Encompass_Preferred
Temperature Monitor Only, Digital, Type "T", 220V, -200 to 250C, with Electrical Plug for China. Temperature monitor features a built in audible alarm, Type T or K thermocouple receptacle, and a bright LED display for easy viewing. The monitor is contained in a compact Stainless Steel cabinet with a side-mounted clamp for securing to a 1/2 diameter support rod. It has a separate ON/OFF switch and a circuit breaker. Dimensions: 2.6 High x 3.2 Wide x 5.5" Deep.
Catalog No. 50-153-1622 Supplier Chemglass Life Sciences Supplier No. CG349801Y
Temperature Monitor Only, Digital, Type "T", 220V, -200 to 250C, with Electrical Plug for China. Temperature monitor features a built in audible alarm, Type T or K thermocouple receptacle, and a bright LED display for easy viewing. The monitor is contained in a compact Stainless Steel cabinet with a side-mounted clamp for securing to a 1/2 diameter support rod. It has a separate ON/OFF switch and a circuit breaker. Dimensions: 2.6 High x 3.2 Wide x 5.5" Deep.