Learn More
Chemglass Life Sciences Temperature Monitor Only, Digital, Type "T", 120V, -200 to 250°C
SDP
Catalog No. CG349801
Encompass_Preferred
Temperature Monitor Only, Digital, Type "T", 120V, -200 to 250¿C. Temperature monitor features a built in audible alarm, Type ¿T¿ or ¿K¿ thermocouple receptacle, and a bright LED display for easy viewing. The monitor is contained in a compact stainless steel cabinet with a side-mounted clamp for securing to a 1/2¿ diameter support rod. It has a separate ON/OFF switch and a circuit breaker. Dimensions: 2.6¿ High x 3.2¿ Wide x 5.5" Deep.PLEASE NOTE: SUPPORT ROD, THERMOCOUPLE AND EXTENSION CORD ARE SOLD SEPARATELY.
Catalog No. CG349801 Supplier Chemglass Life Sciences Supplier No. CG349801
Temperature Monitor Only, Digital, Type "T", 120V, -200 to 250¿C. Temperature monitor features a built in audible alarm, Type ¿T¿ or ¿K¿ thermocouple receptacle, and a bright LED display for easy viewing. The monitor is contained in a compact stainless steel cabinet with a side-mounted clamp for securing to a 1/2¿ diameter support rod. It has a separate ON/OFF switch and a circuit breaker. Dimensions: 2.6¿ High x 3.2¿ Wide x 5.5" Deep.PLEASE NOTE: SUPPORT ROD, THERMOCOUPLE AND EXTENSION CORD ARE SOLD SEPARATELY.