Learn More
Hach Company Digital Termination Box
Supplier:  Hach Company 5867000
Encompass
The Digital Termination box is only used when the desired length of cable between a compatible digital Sensor and the digital Hach SC100 SC200 or SC4500 Controller is between 60 m (197 ft ) and 400 m (1312 ft ) If you do not know if your sensor is compatible with the Digital Termination box please contact Hach Technical Support
Catalog No. 50-002-07250