Learn More
Chemglass Life Sciences Digital Thermometer, Triple Display, with 30mL Bottle, Freezer Applications, -20.0°C Certified Temperature
SDP
Catalog No. 501311345
Encompass
Digital Thermometer, Triple Display, with 30mL Bottle, Freezer Applications, -20.0¿C Certified Temperature. Simultaneously shows the current, minimum, and maximum temperatures and updates continuously. An audible alarm will sound when the temperature rises above or goes below the set temperatures. The minimum/maximum feature monitors critical temperature for any period of time. Reset buttons on front panel. Each thermometer is certified and traceable to NIST and is supplied with a certificate with one critical temperature value calibrated with the reported results. Each thermometer is serialized. The thermometer probe is housed in an unbreakable plastic bottle or vial filled with a buffering media which maintains the thermometer reading when the instrument door is opened. The thermometer and the bottle housing the probe have dual magnets. Other features include a flip-open stand, wall mounting key hole, and supplied with an AAA alkaline battery. The instrument is ¿C and ¿F switchable.
Catalog No. 50-131-1345 Supplier Chemglass Life Sciences Supplier No. CLS408001
Digital Thermometer, Triple Display, with 30mL Bottle, Freezer Applications, -20.0¿C Certified Temperature. Simultaneously shows the current, minimum, and maximum temperatures and updates continuously. An audible alarm will sound when the temperature rises above or goes below the set temperatures. The minimum/maximum feature monitors critical temperature for any period of time. Reset buttons on front panel. Each thermometer is certified and traceable to NIST and is supplied with a certificate with one critical temperature value calibrated with the reported results. Each thermometer is serialized. The thermometer probe is housed in an unbreakable plastic bottle or vial filled with a buffering media which maintains the thermometer reading when the instrument door is opened. The thermometer and the bottle housing the probe have dual magnets. Other features include a flip-open stand, wall mounting key hole, and supplied with an AAA alkaline battery. The instrument is ¿C and ¿F switchable.