Learn More
Cole-Parmer Traceable Digital Thermohygrometer with Dew Point
Supplier:  Cole-Parmer 3780383
Encompass_Preferred
The meter warns you of environmental conditions that are out of a desired range with an audible alarm and bright red LED indicator; alarm may be set up to trigger at three different humidity readings (55%, 60%, or 65%). Features include a super-large, triple display for relative humidity, temperature, and dew point; min/max with reset function; and switchable °F/°C units. Set to always-on feature to monitor conditions for an extended time period; display updates once a second.
Catalog No. NC3789471