Learn More
Cole-Parmer Extech 39240 Digital Waterproof Pocket Thermometer
Supplier:  Cole-Parmer 8648723
Encompass
Reduced Tip Waterproof Thermometer with antimicrobial protection, has a 3'' long stem and a 1.5mm diameter tip. Features °F/°C switchable ranges, a hold function, a display that updates every 1.5 seconds and low battery indicator. Use ON/OFF button to conserve battery life or the unit will automatically shut off after 35 minutes..
Catalog No. 50-993-089