Learn More
Cooper Atkins Corporation DIGITAL THERMOMETER DPP400W08
Catalog No. NC0167656
Encompass_Preferred
This item has a minimum qty of 6 per supplier requirements.
This waterproof digital thermometer is NSF certified and has automatic shut-off. Designed with anti-microbial additive to avoid bacteria growth and reduced stainless steel tip for a quick response time. Temperature Range: -40 to 392F (-40 to 200C).
Catalog No. NC0167656 Supplier Cooper Atkins Corporation Supplier No. DPP400W08
This item has a minimum qty of 6 per supplier requirements.
This waterproof digital thermometer is NSF certified and has automatic shut-off. Designed with anti-microbial additive to avoid bacteria growth and reduced stainless steel tip for a quick response time. Temperature Range: -40 to 392F (-40 to 200C).