Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LaMotte™ Digital Topsoil Tour
SDPSCI_ED

Catalog No. S44609 Shop All LaMotte Company Products
Change view
Click to view available options
Class Size:
50 Students
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S44609 50 Students
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S44609 Supplier LaMotte™ Supplier No. 597301
Add to Cart
Add to Cart

An exciting classroom investigation into the physical and chemical properties of soil.

  • Each student completes seven units while conducting his/her own soil tests for texture, pH, nitrogen, phosphorus and potassium using simple TesTabs™ reagent tablets
  • Incorporates math and science activities, which can be modified to fit schedules and learning opportunities
  • Data sheets and puzzles included
  • Great for inclusion classrooms
  • Bring a soil sample from home, or start a school garden

Specifications

Grade K to 12
Includes Activites, Reagent Tablets, Data Sheets, Puzzles
Class Size 50 Students
Product Type Topsoil Tour Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.