Learn More
BRANSON ULTRASONICS CORPORATION (3P) DIGITAL ULTRASONIC BATH
Catalog No. NC3721484
Encompass_Preferred
Branson Ultrasonics CPX Series Ultrasonic Cleaning Bath, Model: CPX1800, Capacity: 1.9L (0.5 gal.), Dimensions Interior: 15 x 14 x 10cm (6 x 5.5 x 4 in.), Voltage: 120V, Hertz: 40kHz, For Use With: Cleaning, Includes: Tank cover, Plastic, Timer: 90 min., Warranty: 2 Years, Weight: 7 lb., 3.2kg
Catalog No. NC3721484 Supplier BRANSON ULTRASONICS CORPORATION (3P) Supplier No. CPX952119R
Branson Ultrasonics CPX Series Ultrasonic Cleaning Bath, Model: CPX1800, Capacity: 1.9L (0.5 gal.), Dimensions Interior: 15 x 14 x 10cm (6 x 5.5 x 4 in.), Voltage: 120V, Hertz: 40kHz, For Use With: Cleaning, Includes: Tank cover, Plastic, Timer: 90 min., Warranty: 2 Years, Weight: 7 lb., 3.2kg