Learn More
BRANSON ULTRASONICS CORPORATION (3P) DIGITAL ULTRASONIC BATH
Supplier:  BRANSON ULTRASONICS CORPORATION (3P) CPX952319R
Encompass_Preferred
Branson Ultrasonics CPX Series Ultrasonic Cleaning Bath, Model: CPX3800, Capacity: 5.7L (1.5 gal.), Dimensions Interior: 29 x 15 x 15cm (11.5 x 6 x 6 in.), Voltage: 120V, Hertz: 40kHz, For Use With: Cleaning, Includes: Tank cover and drain, Plastic, Timer: 90 min., Warranty: 2 Years
Catalog No. NC3721489