Learn More
BRANSON ULTRASONICS CORPORATION (3P) DIGITAL ULTRASONIC BATH
Catalog No. NC3721491
Encompass_Preferred
Branson Ultrasonics CPX Series Ultrasonic Cleaning Bath, Model: CPX8800, Capacity: 20.8L (5.5 gal.), Dimensions Interior: 49.5 x 29 x 15cm (19.5 x 11.5 x 6 in.), Voltage: 120V, Hertz: 40kHz, For Use With: Cleaning, Includes: Tank cover and drain, Plastic, Timer: 90 min., Warranty: 2 Years
Catalog No. NC3721491 Supplier BRANSON ULTRASONICS CORPORATION (3P) Supplier No. CPX952819R
Branson Ultrasonics CPX Series Ultrasonic Cleaning Bath, Model: CPX8800, Capacity: 20.8L (5.5 gal.), Dimensions Interior: 49.5 x 29 x 15cm (19.5 x 11.5 x 6 in.), Voltage: 120V, Hertz: 40kHz, For Use With: Cleaning, Includes: Tank cover and drain, Plastic, Timer: 90 min., Warranty: 2 Years