Learn More
BRANSON ULTRASONICS CORPORATION (3P) DIGITAL ULTRASONIC BATH
Catalog No. NC3721493
Encompass_Preferred
Branson Ultrasonics CPX Series Ultrasonic Cleaning Bath, Model: CPX5800-E, Capacity: 9.5L (2.5 gal.), Dimensions Interior: 29 x 24 x 15cm (11.5 x 9.5 x 6 in.), Voltage: 230/240V, Hertz: 40kHz, For Use With: Cleaning, Includes: Tank cover and drain, Plastic, Timer: 90 min., Warranty: 2 Years
Catalog No. NC3721493 Supplier BRANSON ULTRASONICS CORPORATION (3P) Supplier No. CPX952539R
Branson Ultrasonics CPX Series Ultrasonic Cleaning Bath, Model: CPX5800-E, Capacity: 9.5L (2.5 gal.), Dimensions Interior: 29 x 24 x 15cm (11.5 x 9.5 x 6 in.), Voltage: 230/240V, Hertz: 40kHz, For Use With: Cleaning, Includes: Tank cover and drain, Plastic, Timer: 90 min., Warranty: 2 Years