Learn More
World Precision Instrument Digital Ultrasonic Cleaner
SDP
Catalog No. 502539358
Encompass
The Ultrasonic Cleaner is a high efficiency cleaner specially developed to clean surgical instruments. The Ultrasonic Cleaner is a quick and effective way to clean lab tools: Ultrasonic waves pump micron-sized bubbles into the water. These bubbles fit into the tightest spaces that would be impossible to clean by traditional methods. When the bubbles implode, water hits the instrument at speeds up to 250 mph.
Catalog No. 50-253-9358 Supplier World Precision Instrument Supplier No. 504495
The Ultrasonic Cleaner is a high efficiency cleaner specially developed to clean surgical instruments. The Ultrasonic Cleaner is a quick and effective way to clean lab tools: Ultrasonic waves pump micron-sized bubbles into the water. These bubbles fit into the tightest spaces that would be impossible to clean by traditional methods. When the bubbles implode, water hits the instrument at speeds up to 250 mph.