Learn More
Thermco Products Inc Triple Display Freezer digital thermometer, Time & Date Stamp, single probe, -50/70 deg.C, 5ml Vaccine bottles probe filled with Bio-Safe medium. Audible & Visual alarms.
Catalog No. NC0601235
Encompass_Preferred
We’re currently building out the product information and will be adding additional content soon. Thank you for your patience as we continue to expand our selection and improve your ordering experience.
Catalog No. NC0601235 Supplier Thermco Products Inc Supplier No. ACC821FREV