Learn More
Thermco Products Inc Triple Display Refrigerator digital thermometer, Time & Date Stamp, single probe, -50/70 deg.C, 5ml Vaccine bottles probe filled with Bio-Safe medium. Audible & Visual alarms.
Catalog No. NC0601236
Encompass_Preferred
Triple Display Digital Thermometer simultaneously shows the current, minimum, and maximum temperatures and updates continuously. An audible & visual alarm will trigger when the temperatures goes above or below the set temperatures. The minimum/maximum feature monitors critical temperature for any period of time, such as overnight or weekends. To comply with regulatory agencies requirements this Time-Stamp thermometer provides the date and time when the temperature being monitored exceeds the minimum and maximum settings and in real-time. Also an audible alarm will sound and flashing LED will be visible.
Catalog No. NC0601236 Supplier Thermco Products Inc Supplier No. ACC821REFV
Triple Display Digital Thermometer simultaneously shows the current, minimum, and maximum temperatures and updates continuously. An audible & visual alarm will trigger when the temperatures goes above or below the set temperatures. The minimum/maximum feature monitors critical temperature for any period of time, such as overnight or weekends. To comply with regulatory agencies requirements this Time-Stamp thermometer provides the date and time when the temperature being monitored exceeds the minimum and maximum settings and in real-time. Also an audible alarm will sound and flashing LED will be visible.