Learn More
Digivac Model 200m | Laboratory Grade Vacuum Controller (Microns)
SDP
Catalog No. NC0934392
Encompass_Preferred
A variation of DigiVac’s best-selling Model 200 with readings in microns. This digital vacuum controller and gauge measures through 6 decades of wide range rough vacuum with precise accuracy. ◦The gauge controller is housed in a laboratory grade case consisting of vinyl clad metal on an extruded aluminum base insuring maximum durability◦This kit comes with everything to measure vacuum: Display controller, cables, power and 531 thermocouple sensor
Catalog No. NC0934392 Supplier Digivac Supplier No. M200
Add to Cart
Add to Cart
A variation of DigiVac’s best-selling Model 200 with readings in microns. This digital vacuum controller and gauge measures through 6 decades of wide range rough vacuum with precise accuracy. ◦The gauge controller is housed in a laboratory grade case consisting of vinyl clad metal on an extruded aluminum base insuring maximum durability◦This kit comes with everything to measure vacuum: Display controller, cables, power and 531 thermocouple sensor