Learn More
Electron Microscopy Sciences Digital Video Color Camera USB 2.0, 5 MP, Stereo
SDP
Supplier:  Electron Microscopy Sciences 575M
Encompass_Preferred
Each microscope camera system is fully integrated with software for image analysis, optical adapters to obtain the image in the correct focal plane, and cables for connection to computers, monitors or projectors. USB 2.0, 5 Megapixel resolution for color imaging, optical adapter and measuring software.
Type: Stereo
Catalog No. 50-190-2316