Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OHAUS Digital Vortex Genie 2

Catalog No. 01804411 Shop All OHAUS Products
Change view
Click to view available options
Product Type:
Vortex
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
01-804-411 Vortex
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-804-411 Supplier OHAUS Supplier No. 83041322
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Digital control and display of speed and time.

  • Ideal for repeat applications.
  • Digital timer from 1 to 99 sec. for Touch and 1 to 99 min. for Hands-Free operations.
  • Accepts all Vortex Genie 2 attachments.
TRUSTED_SUSTAINABILITY

Specifications

Orbit 1 mm
Speed Range 500 to 3000 rpm
Voltage 120 V
Product Type Vortex
Model Digital Vortex-Genie 2
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.