Learn More
Electron Microscopy Sciences Digital Vortex Genie 2 European Plug, 120 V
SDP
Catalog No. 501899677
Encompass_Preferred
Digital Vortex-Genie 2 with variable speed control allows slow speed shaking action up to high-speed vortexing. Each optional attachment is designed to produce different mixing actions, allowing users to choose the correct action fo the tube or container. Touch on for 1-99 seconds and hands-free for 1-99 minutes or continuous with timer function. Tough metal housing combined with just enough weight provides a stable platform for all types of mixing.
Catalog No. 50-189-9677 Supplier Electron Microscopy Sciences Supplier No. 71838EU
Digital Vortex-Genie 2 with variable speed control allows slow speed shaking action up to high-speed vortexing. Each optional attachment is designed to produce different mixing actions, allowing users to choose the correct action fo the tube or container. Touch on for 1-99 seconds and hands-free for 1-99 minutes or continuous with timer function. Tough metal housing combined with just enough weight provides a stable platform for all types of mixing.