Learn More
MSC Supco Digital Wall Thermostat, Single Stage, Non-Programmable, Backlit, 1 ea
Catalog No. 5000509517
Encompass
This single-stage digital wall thermostat provides electronic temperature control for conventional and single-stage HVAC systems, including gas, oil, and electric furnaces, as well as single-stage heat pumps. Designed for simple, manual operation, the unit features a large, blue-backlit LCD for clear visibility and consistent temperature regulation.
  • Compatible with gas, oil, electric, and single-stage heat pump systems
  • Features a blue-backlit LCD for easy reading in low light
  • Includes a 3.5 to 4 minute compressor protection cycle delay
  • Includes an INDIGLO® night-light that activates with key presses
  • Operating temperature range of 45°F to 95°F
  • Non-programmable for manual operation
  • Includes front-access battery compartment
  • Automatic system cutoff in heat and cool modes
  • Includes 2 AA alkaline batteries
Catalog No. 50-005-09517 Supplier MSC Supplier No. 91965277
This single-stage digital wall thermostat provides electronic temperature control for conventional and single-stage HVAC systems, including gas, oil, and electric furnaces, as well as single-stage heat pumps. Designed for simple, manual operation, the unit features a large, blue-backlit LCD for clear visibility and consistent temperature regulation.
  • Compatible with gas, oil, electric, and single-stage heat pump systems
  • Features a blue-backlit LCD for easy reading in low light
  • Includes a 3.5 to 4 minute compressor protection cycle delay
  • Includes an INDIGLO® night-light that activates with key presses
  • Operating temperature range of 45°F to 95°F
  • Non-programmable for manual operation
  • Includes front-access battery compartment
  • Automatic system cutoff in heat and cool modes
  • Includes 2 AA alkaline batteries