Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mopec™ Digital Washdown Platform Scale
SDP

Catalog No. 22445641 Shop All Mopec Products
Change view
Click to view available options
Capacity:
15,000 x 5 g (300 x 0.1 oz.)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity
22-445-641 15,000 x 5 g (300 x 0.1 oz.)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-445-641 Supplier Mopec™ Supplier No. BB066
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The washdown base provides a durable and high performance weighing package equipped with a removable stainless cover to enable easy wipe down of the all-stainless exterior.

  • Compact and easy to use
  • IP68 water and dust resistant
  • Stainless steel finish
  • Removable weigh pan easy cleaning
  • LED display
  • AC adapter included

Specifications

Capacity 15,000 x 5 g (300 x 0.1 oz.)
Readability 30 kg x 10 g
Display Type Digital
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.