Learn More
Yamato Scientific America, Inc. Digital Constant Temperature Water Bath 5L 100-115V
Catalog No. NC1769325 Shop All Yamato Scientific America Inc. Products
Encompass_Preferred
Large capacity 5L constant temperature water bath with internal diameter of φ240 mm. Designed with flat-shaped bath with no heater or sensor inside for ease in cleaning. Temperature range: ambient +10~90°C. 100-115V.
Catalog No. NC1769325 Supplier Yamato Scientific America, Inc. Supplier No. BM302A
Large capacity 5L constant temperature water bath with internal diameter of φ240 mm. Designed with flat-shaped bath with no heater or sensor inside for ease in cleaning. Temperature range: ambient +10~90°C. 100-115V.