Learn More
Yamato Scientific America, Inc. Digital Constant Temperature Water Bath 5L 200-230V
Catalog No. NC1769326 Shop All Yamato Scientific America Inc. Products
Encompass_Preferred
Large capacity 5L constant temperature water bath with internal diameter of φ240 mm. Designed with flat-shaped bath with no heater or sensor inside for ease in cleaning. Temperature range: ambient +10~90°C. 200-230V.
Catalog No. NC1769326 Supplier Yamato Scientific America, Inc. Supplier No. BM312A
Large capacity 5L constant temperature water bath with internal diameter of φ240 mm. Designed with flat-shaped bath with no heater or sensor inside for ease in cleaning. Temperature range: ambient +10~90°C. 200-230V.