Learn More
Research Products International Corp Digital Control Water Bath with Lid, 6 Liter Capacity, 230V European Plug
SDP
Catalog No. 501367892
Encompass
Hot Tubs Water Baths offer a variety of sizes to meet all of your needs and are ideal for general lab applications. The space saving design allows for the most efficient use of bench space and the PID Controller offers easy temperature selection, rapid heat-up and excellent stability. Baths can also be used as a boiling water bath. User simply sets temperature to “boiling mode” and attaches gabled cover.
Large easy to read LED panel displays temperature settings in 0.1°C increments and programmable timer up to 99hrs, 59mins. Temperatures from +5°C above ambient up to 100°C can be maintained with ±0.1°C accuracy. Recessed panel protects controls from spills. Seamless steel tank includes diffuser tray and shelf. Built for years of reliable performance and include a two year warranty.
Excellent performance
Boiling mode is ideal for melting agar
Two year warranty
Catalog No. 50-136-7892 Supplier Research Products International Corp Supplier No. 246600230
Hot Tubs Water Baths offer a variety of sizes to meet all of your needs and are ideal for general lab applications. The space saving design allows for the most efficient use of bench space and the PID Controller offers easy temperature selection, rapid heat-up and excellent stability. Baths can also be used as a boiling water bath. User simply sets temperature to “boiling mode” and attaches gabled cover.
Large easy to read LED panel displays temperature settings in 0.1°C increments and programmable timer up to 99hrs, 59mins. Temperatures from +5°C above ambient up to 100°C can be maintained with ±0.1°C accuracy. Recessed panel protects controls from spills. Seamless steel tank includes diffuser tray and shelf. Built for years of reliable performance and include a two year warranty.
Excellent performance
Boiling mode is ideal for melting agar
Two year warranty