Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LaMotte™ Digital Watershed Tour
SDPSCI_ED

Catalog No. S44605 Shop All LaMotte Company Products
Change view
Click to view available options
Class Size:
50 Students (8 Groups)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S44605 50 Students (8 Groups)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S44605 Supplier LaMotte™ Supplier No. 5974
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mini digital curriculum to introduce students to watersheds virtually in the classroom.

  • Introduce students to stream and water ecosystems through a classroom-based exploration of the virtual Big River watershed
  • Students learn about stream ecology, water quality and how their own actions can affect a watershed
  • Through hands-on activities, student groups evaluate simulated water samples to collect chemical and biological data from four mystery sites
  • Students apply their knowledge of watersheds to analyze the data and identify their site
  • Groups write an action plan for the Big River river keeper for improving or maintaining water quality
  • Activities can be modified to fit schedules and learning opportunities
  • Data sheets and puzzles included
  • Great for inclusion classrooms

Specifications

Grade K to 12
Includes Virtual Tour, Data Sheets, Puzzles
Class Size 50 Students (8 Groups)
Product Type Watershed Tour Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.