Learn More
Cayman Chemical DIgItals purpurea DIgItoIn 1g
SDP
Catalog No. 502971133
Encompass_Preferred
A non-ionic detergent used to solubilize membrane-bound proteins, to precipitate cholesterol, and to permeabilize cell membranes
Catalog No. 50-297-1133 Supplier Cayman Chemical Supplier No. 149521 G
A non-ionic detergent used to solubilize membrane-bound proteins, to precipitate cholesterol, and to permeabilize cell membranes