Learn More
Cayman Chemical DIgItls pururea DIgItIn 100mg
SDP
Catalog No. 502972744
Encompass_Preferred
A non-ionic detergent used to solubilize membrane-bound proteins, to precipitate cholesterol, and to permeabilize cell membranes
Catalog No. 50-297-2744 Supplier Cayman Chemical Supplier No. 14952100 MG
A non-ionic detergent used to solubilize membrane-bound proteins, to precipitate cholesterol, and to permeabilize cell membranes