Learn More
Research Products International Corp Digitonin, Water Soluble, 1 Gram
SDP
Catalog No. 50488646
Encompass_Preferred
Non-ionic detergent used to solubilize membrane bound proteins.
Catalog No. 50-488-646 Supplier Research Products International Corp Supplier No. D430651.0
Non-ionic detergent used to solubilize membrane bound proteins.