Learn More
Research Products International Corp Digitonin, Water Soluble, 100 Milligrams
SDP
Catalog No. 50488644
Encompass_Preferred
Non-ionic detergent used to solubilize membrane bound proteins.
Catalog No. 50-488-644 Supplier Research Products International Corp Supplier No. D430650.1
Non-ionic detergent used to solubilize membrane bound proteins.