Learn More
Medchemexpress LLC Digitonin | 11024-24-1 | 500 MG

Supplier: Medchemexpress LLC HYN4000500MG
Digitonin is a glycoside derived from Digitalis purpurea. Functioning as a natural detergent, it can enhance cell permeability by binding to cholesterol molecules and has shown potential to reduce tumor growth. This product is for research use only.
- Obtained from Digitalis purpurea
- Increases cell permeability by binding to cholesterol molecules
- May reduce tumor growth
- Natural detergent properties
- Purity of 54.4%
- Solid appearance, off-white to light yellow color
- Store at 4°C, under nitrogen for solid form
- Soluble in water at 100 mg/mL (requires ultrasonic)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.