Learn More
Medchemexpress LLC HY-N4000 25mg Medchemexpress, Digitonin CAS:11024-24-1 Purity:>98%
SDP
Catalog No. 501933941
Encompass_Preferred
Medchemexpress, HY-N4000 25mg Digitonin CAS:11024-24-1 Digitonin, a glycoside obtained from Digitalis purpurea, could increase cell permeability by binding to cholesterol molecules and reduce tumor growth. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3941 Supplier Medchemexpress LLC Supplier No. HYN400025MG
Medchemexpress, HY-N4000 25mg Digitonin CAS:11024-24-1 Digitonin, a glycoside obtained from Digitalis purpurea, could increase cell permeability by binding to cholesterol molecules and reduce tumor growth. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.