Learn More
Selleck Chemical LLC Digitonin-E1321-25MG
SDP
Catalog No. 502390053
Encompass_Preferred
Digitonin a steroidal saponin and glycoside obtained from Digitalis purpurea has strong lytic activity on various cell membranes with an IC50 of 15 1 M on sheep RBC membranes
Catalog No. 50-239-0053 Supplier Selleck Chemical LLC Supplier No. E132125MG
Digitonin a steroidal saponin and glycoside obtained from Digitalis purpurea has strong lytic activity on various cell membranes with an IC50 of 15 1 M on sheep RBC membranes