Learn More
FUJIFILM BIOSCIENCES INC Digitonin
Catalog No. 5000450207
Encompass_Preferred
Digitonin is a spirostan saponin obtained from the seeds and leaves of digitalis D purpurea D lanata A mild nonionic detergent digitonin is used in a wide range of applications including solubilization of membrane proteins permeabilization of cellular membranes isolation of mitochondria and quantitation of cholesterol Water Soluble 4% For research use only RUO
Catalog No. 50-004-50207 Supplier FUJIFILM BIOSCIENCES INC Supplier No. 04721374
Add to Cart
Add to Cart
Digitonin is a spirostan saponin obtained from the seeds and leaves of digitalis D purpurea D lanata A mild nonionic detergent digitonin is used in a wide range of applications including solubilization of membrane proteins permeabilization of cellular membranes isolation of mitochondria and quantitation of cholesterol Water Soluble 4% For research use only RUO