Learn More
Sigma Aldrich Fine Chemicals Biosciences Digitonin, 11024-24-1, MFCD00077729, 5 g
Catalog No. NC1106101
Encompass
Empirical Formula (Hill Notation): C56H92O29, Molecular Weight: 1229.31, ~50% (TLC), mp: 230-240 °C, Synonym: Digitin.
Catalog No. NC1106101 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. D56285G
Empirical Formula (Hill Notation): C56H92O29, Molecular Weight: 1229.31, ~50% (TLC), mp: 230-240 °C, Synonym: Digitin.