Learn More
Crescent Chemical Co Inc DIGITONIN ANALYTICAL GRADE
SDP
Catalog No. 50247512
Encompass_Preferred
Digitonin analytical grade, USP Naturally occurring surfactant especially useful for receptor solubilization. Frequently applied as hemolysis reagent and for permeabilization of certain types of cells e.g. blood platelets, hepatocytes, yeast or tumor cells.
Catalog No. 50-247-512 Supplier Crescent Chemical Co Inc Supplier No. SE19550.02
Digitonin analytical grade, USP Naturally occurring surfactant especially useful for receptor solubilization. Frequently applied as hemolysis reagent and for permeabilization of certain types of cells e.g. blood platelets, hepatocytes, yeast or tumor cells.